1-Chloro-4-(chlorodifluoromethanesulphonyl)benzene
Lieferant: Apollo Scientific
PC10696-1G
PC10696-250MG
APOSPC10696-1GEA
719
EUR
APOSPC10696-1G
APOSPC10696-250MG
1-Chloro-4-(chlorodifluoromethanesulphonyl)benzene
1-Chloro-4-(chlorodifluoromethanesulphonyl)benzene
Formel:
C₇H₄Cl₂F₂O₂S Molecular Weight: 261,073 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11500247 CAS-Nummer: 403496-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...