6-Chloro-5-fluoro-2-(pyrazin-2-yl)-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(6-Chloro-5-fluoro-1H-benzimidazol-2-yl)pyrazine

PC10701-1G PC10701-5G
APOSPC10701-1GEA 139 EUR
APOSPC10701-1G APOSPC10701-5G
6-Chloro-5-fluoro-2-(pyrazin-2-yl)-1H-benzimidazole
6-Chloro-5-fluoro-2-(pyrazin-2-yl)-1H-benzimidazole
Formel: C₁₁H₆ClFN₄
Molecular Weight: 248,644 g/mol
Schmelzpunkt: 284…287 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16606068
CAS-Nummer: 1257535-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR