3-Bromo-2-fluoro-6-iodonitrobenzene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC10729-1G
PC10729-250MG
APOSPC10729-1GEA
654
EUR
APOSPC10729-1G
APOSPC10729-250MG
3-Bromo-2-fluoro-6-iodonitrobenzene 95%
3-Bromo-2-fluoro-6-iodonitrobenzene
Formel:
C₆H₂BrFINO₂ Molecular Weight: 345,892 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28739556 CAS-Nummer: 1807173-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...