3-Amino-5-fluorobenzo[b]thiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC110002-1G
PC110002-250MG
APOSPC110002-1GEA
188
EUR
APOSPC110002-1G
APOSPC110002-250MG
3-Amino-5-fluorobenzo[b]thiophene-2-carboxylic acid
3-Amino-5-fluorobenzo[b]thiophene-2-carboxylic acid
Formel:
C₉H₆FNO₂S Molecular Weight: 211,213 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12407357 CAS-Nummer: 1378864-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...