3-Amino-6-fluorobenzo[b]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC110003-1G PC110003-250MG
APOSPC110003-1GEA 499 EUR
APOSPC110003-1G APOSPC110003-250MG
3-Amino-6-fluorobenzo[b]thiophene-2-carboxylic acid
3-Amino-6-fluorobenzo[b]thiophene-2-carboxylic acid
Formel: C₉H₆FNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD24496328
CAS-Nummer: 452910-20-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR