3-Amino-7-fluorobenzo[b]thiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC110004-1G
PC110004-250MG
APOSPC110004-1GEA
499
EUR
APOSPC110004-1G
APOSPC110004-250MG
3-Amino-7-fluorobenzo[b]thiophene-2-carboxylic acid
3-Amino-7-fluorobenzo[b]thiophene-2-carboxylic acid
Formel:
C₉H₆FNO₂S Molecular Weight: 211.213 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28606529 CAS-Nummer: 1552532-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...