7-Bromo-4-chloro-8-fluoroquinoline

Lieferant: Apollo Scientific
Achtung

PC110007-1G PC110007-250MG
APOSPC110007-1GEA 473 EUR
APOSPC110007-1G APOSPC110007-250MG
7-Bromo-4-chloro-8-fluoroquinoline
7-Bromo-4-chloro-8-fluoroquinoline
Formel: C₉H₄BrClFN
Molecular Weight: 260.49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32200320
CAS-Nummer: 2111439-88-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR