7-Bromo-8-fluoroquinolin-4-ol

Lieferant: Apollo Scientific
Achtung

PC110009-1G PC110009-250MG
APOSPC110009-1GEA 277 EUR
APOSPC110009-1G APOSPC110009-250MG
7-Bromo-8-fluoroquinolin-4-ol
7-Bromo-8-fluoroquinolin-4-ol
Formel: C₉H₅BrFNO
Molecular Weight: 242,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32200321
CAS-Nummer: 2111860-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR