3-Amino-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC110015-1G PC110015-250MG
APOSPC110015-1GEA 208 EUR
APOSPC110015-1G APOSPC110015-250MG
3-Amino-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
3-Amino-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
Formel: C₁₀H₆F₃NO₂S
Molecular Weight: 261.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29169131
CAS-Nummer: 1609255-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR