3-Amino-7-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC110017-1G PC110017-250MG
APOSPC110017-1GEA 328 EUR
APOSPC110017-1G APOSPC110017-250MG
3-Amino-7-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
3-Amino-7-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
Formel: C₁₀H₆F₃NO₂S
Molecular Weight: 261,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29169138
CAS-Nummer: 1648807-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR