3-Amino-2-(trifluoromethyl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC110022-1G
PC110022-250MG
APOSPC110022-1GEA
575
EUR
APOSPC110022-1G
APOSPC110022-250MG
3-Amino-2-(trifluoromethyl)benzoic acid
3-Amino-2-(trifluoromethyl)benzoic acid
Formel:
C₈H₆F₃NO₂ Molecular Weight: 205.134 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22480979 CAS-Nummer: 1369896-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...