6-Cyano-3-fluoropyridin-2-yl acetate

Lieferant: Apollo Scientific
Gefahr

PC110025-1G PC110025-250MG
APOSPC110025-1GEA 719 EUR
APOSPC110025-1G APOSPC110025-250MG
6-Cyano-3-fluoropyridin-2-yl acetate
6-Cyano-3-fluoropyridin-2-yl acetate
Formel: C₈H₅FN₂O₂
Molecular Weight: 180.136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19690162
CAS-Nummer: 1189757-54-7
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR