Methyl 3-amino-6-fluorobenzo[b]thiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC110026-1G PC110026-250MG
APOSPC110026-1GEA 319 EUR
APOSPC110026-1G APOSPC110026-250MG
Methyl 3-amino-6-fluorobenzo[b]thiophene-2-carboxylate
Methyl 3-amino-6-fluorobenzo[b]thiophene-2-carboxylate
Formel: C₁₀H₈FNO₂S
Molecular Weight: 225,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08706231
CAS-Nummer: 142363-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR