Methyl 3-amino-7-fluorobenzo[b]thiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC110027-1G PC110027-250MG
APOSPC110027-1GEA 319 EUR
APOSPC110027-1G APOSPC110027-250MG
Methyl 3-amino-7-fluorobenzo[b]thiophene-2-carboxylate
Methyl 3-amino-7-fluorobenzo[b]thiophene-2-carboxylate
Formel: C₁₀H₈FNO₂S
Molecular Weight: 225,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29158206
CAS-Nummer: 2090172-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR