2-Amino-4,5-difluorobenzonitrile

Lieferant: Apollo Scientific

PC110108-1G PC110108-5G PC110108-500MG
APOSPC110108-1GEA 183 EUR
APOSPC110108-1G APOSPC110108-5G APOSPC110108-500MG
2-Amino-4,5-difluorobenzonitrile
2-Amino-4,5-difluorobenzonitrile
Formel: C₇H₄F₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09835282
CAS-Nummer: 219823-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR