5-Amino-2,4-difluorobenzonitrile

Lieferant: Apollo Scientific

PC110109-1G PC110109-5G
APOSPC110109-1GEA 328 EUR
APOSPC110109-1G APOSPC110109-5G
5-Amino-2,4-difluorobenzonitrile
5-Amino-2,4-difluorobenzonitrile
Formel: C₇H₄F₂N₂
Molecular Weight: 154,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756744
CAS-Nummer: 952285-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR