3-Amino-2,5-difluorobenzonitrile

Lieferant: Apollo Scientific

PC110111-1G PC110111-5G
APOSPC110111-1GEA 509 EUR
APOSPC110111-1G APOSPC110111-5G
3-Amino-2,5-difluorobenzonitrile
3-Amino-2,5-difluorobenzonitrile
Formel: C₇H₄F₂N₂
Molecular Weight: 154,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28059689
CAS-Nummer: 1805663-68-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR