3-[4-(3-(Trifluoromethyl)phenoxy)phenyl]-1H-pyrazole

Lieferant: Apollo Scientific

PC110116-1G PC110116-250MG
APOSPC110116-1GEA 261 EUR
APOSPC110116-1G APOSPC110116-250MG
3-[4-(3-(Trifluoromethyl)phenoxy)phenyl]-1H-pyrazole
3-[4-(3-(Trifluoromethyl)phenoxy)phenyl]-1H-pyrazole
Formel: C₁₆H₁₁F₃N₂O
Molecular Weight: 304.267 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06409163
CAS-Nummer: 1019007-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR