Tris(ethyl)amine pentahydrofluoride

Lieferant: Apollo Scientific
Gefahr

PC11105-25G PC11105-5G
APOSPC11105-25GEA 139 EUR
APOSPC11105-25G APOSPC11105-5G
Tris(ethyl)amine pentahydrofluoride
Tris(ethyl)amine pentahydrofluoride
Formel: (FH)₅·(C₂H₅)₃N
Molecular Weight: 201,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797182
CAS-Nummer: 94527-74-9
UN: 2927
ADR: 6.1+8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR