3-Amino-4'-(trifluoromethyl)biphenyl hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-(3-Aminophenyl)benzotrifluoride hydrochloride, 3-[4-(Trifluoromethyl)phenyl]aniline hydrochloride, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine hydrochloride

PC11164-1G PC11164-250MG
APOSPC11164-1GEA 504 EUR
APOSPC11164-1G APOSPC11164-250MG
3-Amino-4'-(trifluoromethyl)biphenyl hydrochloride
3-Amino-4'-(trifluoromethyl)biphenyl hydrochloride
Formel: C₁₃H₁₀F₃N·ClH
Molecular Weight: 273.681 g/mol
MDL: MFCD08445323
CAS-Nummer: 1211195-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR