3-Bromo-4-chloro-2-(trifluoromethyl)quinoline

Lieferant: Apollo Scientific
Achtung

PC11259-1G PC11259-2.5G
APOSPC11259-1GEA 251 EUR
APOSPC11259-1G APOSPC11259-2.5G
3-Bromo-4-chloro-2-(trifluoromethyl)quinoline
3-Bromo-4-chloro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₄BrClF₃N
Molecular Weight: 310,498 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407379
CAS-Nummer: 683274-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR