2,6-Difluoro-3-hydroxybenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-2,4-difluorophenol, 2,6 Difluoro 3-hydroxy benzoic acid

PC1133-1G PC1133-250MG
APOSPC1133-1GEA 29 EUR
APOSPC1133-1G APOSPC1133-250MG
2,6-Difluoro-3-hydroxybenzoic acid
2,6-Difluoro-3-hydroxybenzoic acid
Formel: C₇H₄F₂O₃
Siedepunkt: 110 °C (19 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD08275286
CAS-Nummer: 749230-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR