5-(4-Fluorophenyl)-1,3,4-oxadiazole-2-thiol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Fluorophenyl)-5-mercapto-1,3,4-oxadiazole, 2-(4-Fluorophenyl)-5-sulphanyl-1,3,4-oxadiazole
PC1198-1G
PC1198-25G
PC1198-5G
APOSPC1198-1GEA
151
EUR
APOSPC1198-1G
APOSPC1198-25G
APOSPC1198-5G
5-(4-Fluorophenyl)-1,3,4-oxadiazole-2-thiol
5-(4-Fluorophenyl)-1,3,4-oxadiazole-2-thiol
Formel:
C₈H₅FN₂OS Molecular Weight: 196,202 g/mol Schmelzpunkt: 176…178 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03197058 CAS-Nummer: 203268-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...