3-Cyano-2,6-dihydroxy-4-(trifluoromethyl)pyridine dihydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dihydroxy-4-(trifluoromethyl)nicontinonitrile dihydrate

PC1219-1G PC1219-25G PC1219-5G
APOSPC1219-1GEA 21.9 EUR
APOSPC1219-1G APOSPC1219-25G APOSPC1219-5G
3-Cyano-2,6-dihydroxy-4-(trifluoromethyl)pyridine dihydrate
3-Cyano-2,6-dihydroxy-4-(trifluoromethyl)pyridine dihydrate
Formel: C₇H₃F₃N₂O₂·2H₂O
Molecular Weight: 240,137 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368671
CAS-Nummer: 1049729-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR