1-[4-(Trifluoromethoxy)phenyl]ethylamine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-(Trifluoromethoxy)phenyl)ethanamine
PC1249-1G
PC1249-50MG
PC1249-5G
APOSPC1249-1GEA
260
EUR
APOSPC1249-1G
APOSPC1249-50MG
APOSPC1249-5G
1-[4-(Trifluoromethoxy)phenyl]ethylamine 98%
1-[4-(Trifluoromethoxy)phenyl]ethylamine
Formel:
C₉H₁₀F₃NO Molecular Weight: 205,177 g/mol Siedepunkt: 69,7…70.1 °C (6.1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784301 CAS-Nummer: 123195-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...