1,1,1,2-Tetrafluoropropane (FC-254eb) 97%

Lieferant: Apollo Scientific
Gefahr

PC1331-25G PC1331-5G
APOSPC1331-25GEA 757 EUR
APOSPC1331-25G APOSPC1331-5G
1,1,1,2-Tetrafluoropropane (FC-254eb) 97%
1,1,1,2-Tetrafluoropropane (FC-254eb)
Formel: C₃H₄F₄
Molecular Weight: 116,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09998069
CAS-Nummer: 421-48-7
UN: 3161
ADR: 2.1,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR