3,4,5,6-Tetrafluorophthalimide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1,3-Dioxo-4,5,6,7-tetrafluoroisoindoline, 4,5,6,7-Tetrafluoro-1H-isoindole-1,3(2H)-dione, 4,5,6,7-Tetrafluoroisoindoline-1,3-dione

PC1405-1G PC1405-25G PC1405-5G
APOSPC1405-1GEA 94.4 EUR
APOSPC1405-1G APOSPC1405-25G APOSPC1405-5G
3,4,5,6-Tetrafluorophthalimide
3,4,5,6-Tetrafluorophthalimide
Formel: C₈HF₄NO₂
Molecular Weight: 219,093 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00091519
CAS-Nummer: 652-11-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR