3',5'-Dichloro-2'-fluoroacetophenone
Lieferant: Apollo Scientific
Synonyme:
1-(3,5-Dichloro-2-fluorophenyl)ethanone
PC1433-5G
APOSPC1433-5GEA
59.6
EUR
APOSPC1433-5G
3',5'-Dichloro-2'-fluoroacetophenone
3',5'-Dichloro-2'-fluoroacetophenone
Formel:
C₈H₅Cl₂FO Molecular Weight: 207,029 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427276 CAS-Nummer: 480438-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...