3,5-Bis(trifluoromethyl)-2,6-dichlorobenzal chloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,5-Bis(trifluoromethyl)-2,4-dichloro-3-(dichloromethyl)benzene

PC1464-1G PC1464-5G
APOSPC1464-1GEA 92.8 EUR
APOSPC1464-1G APOSPC1464-5G
3,5-Bis(trifluoromethyl)-2,6-dichlorobenzal chloride
3,5-Bis(trifluoromethyl)-2,6-dichlorobenzal chloride
Formel: C₉H₂Cl₄F₆
Lagertemperatur: Raumtemperatur
MDL: MFCD03094129
CAS-Nummer: 501657-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR