3-Fluorophenylacetyl chloride 97+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(3-Fluorophenyl)ethanoyl chloride
PC1582-25G
PC1582-5G
PC1582-100G
APOSPC1582-25GEA
140
EUR
APOSPC1582-25G
APOSPC1582-5G
APOSPC1582-100G
3-Fluorophenylacetyl chloride 97+%
3-Fluorophenylacetyl chloride
Formel:
C₈H₆ClFO Molecular Weight: 172,584 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787556 CAS-Nummer: 458-04-8 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...