2'-Bromo-4'-(trifluoromethyl)acetanilide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Acetamido-3-bromobenzotrifluoride, N-[2-Bromo-4-(trifluoromethyl)phenyl]acetamide

PC1593Y-10G PC1593Y-1G PC1593Y-5G
APOSPC1593Y-10GEA 92.8 EUR
APOSPC1593Y-10G APOSPC1593Y-1G APOSPC1593Y-5G
2'-Bromo-4'-(trifluoromethyl)acetanilide 97%
2'-Bromo-4'-(trifluoromethyl)acetanilide
Formel: C₉H₇BrF₃NO
Molecular Weight: 282,057 g/mol
Schmelzpunkt: 144…146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067782
CAS-Nummer: 175135-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR