3-(Difluoromethoxy)benzeneboronic acid 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Borono-3-(difluoromethoxy)benzene, (3-(Difluoromethoxy)phenyl)boronic acid

PC1596-1G PC1596-10G PC1596-5G PC1596-25G
APOSPC1596-1GEA 85.7 EUR
APOSPC1596-1G APOSPC1596-10G APOSPC1596-5G APOSPC1596-25G
3-(Difluoromethoxy)benzeneboronic acid 99%
3-(Difluoromethoxy)benzeneboronic acid
Formel: C₇H₇BF₂O₃
Molecular Weight: 187,936 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08706282
CAS-Nummer: 866607-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR