3-(Difluoromethyl)benzeneboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-(Difluoromethyl)phenyl)boronic acid
PC1641-1G
APOSPC1641-1GEA
109
EUR
APOSPC1641-1G
3-(Difluoromethyl)benzeneboronic acid
3-(Difluoromethyl)benzeneboronic acid
Formel:
C₇H₇BF₂O₂ Molecular Weight: 171,937 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09864670 CAS-Nummer: 854690-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...