3-Fluoro-4-(hydrazinocarbonyl)benzeneboronic acid 96%
Lieferant: Apollo Scientific
Synonyme:
3-Fluoro-4-hydrazinocarbonylphenylboronic acid
PC1663-1G
PC1663-5G
APOSPC1663-1GEA
147
EUR
APOSPC1663-1G
APOSPC1663-5G
3-Fluoro-4-(hydrazinocarbonyl)benzeneboronic acid 96%
3-Fluoro-4-(hydrazinocarbonyl)benzeneboronic acid
Formel:
C₇H₈BFN₂O₃ Molecular Weight: 197,959 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973743 CAS-Nummer: 850568-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...