2-(Aminomethyl)-4-fluorobenzeneboronic acid, N-BOC protected 97%
Lieferant: Apollo Scientific
Achtung
PC1681-1G PC1681-5G
APOSPC1681-1GEA 229 EUR
APOSPC1681-1G APOSPC1681-5G
2-(Aminomethyl)-4-fluorobenzeneboronic acid, N-BOC protected 97%
2-(Aminomethyl)-4-fluorobenzeneboronic acid, N-BOC protected
Formel: C₁₂H₁₇BFNO₄
Molecular Weight: 269.077 g/mol
Schmelzpunkt: 90…94 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659826
CAS-Nummer: 850568-64-8