2,2-Dimethyl-4,4,4-trifluorobutanoic acid 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,2-Dimethyl-4,4,4-trifluorobutyric acid

PC1824-1G PC1824-250MG
APOSPC1824-1GEA 613 EUR
APOSPC1824-1G APOSPC1824-250MG
2,2-Dimethyl-4,4,4-trifluorobutanoic acid 95%
2,2-Dimethyl-4,4,4-trifluorobutanoic acid
Formel: C₆H₉F₃O₂
Molecular Weight: 170,13 g/mol
Siedepunkt: 86…87 °C (6 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD09998070
CAS-Nummer: 939399-07-2
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR