3-Chloro-2,4,5-trifluorobenzoyl chloride 98%

Lieferant: Apollo Scientific
Gefahr

PC1853-10G PC1853-25G PC1853-100G
APOSPC1853-10GEA 81.2 EUR
APOSPC1853-10G APOSPC1853-25G APOSPC1853-100G
3-Chloro-2,4,5-trifluorobenzoyl chloride 98%
3-Chloro-2,4,5-trifluorobenzoyl chloride
Formel: C₇HCl₂F₃O
Molecular Weight: 228,983 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039245
CAS-Nummer: 101513-78-4
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR