2-(4-Chlorophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-α,α-bis(trifluoromethyl)benzyl alcohol, Bis(trifluoromethyl)(4-chlorophenyl)carbinol, 1-Chloro-4-[perfluoro(2-hydroxyprop-2-yl)]benzene

PC1931B-1G
APOSPC1931B-1GEA 109 EUR
APOSPC1931B-1G
2-(4-Chlorophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 98%
2-(4-Chlorophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Formel: C₉H₅ClF₆O
Molecular Weight: 278,579 g/mol
Siedepunkt: 189…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041489
CAS-Nummer: 2010-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR