3-Fluoro-2-hydrazinopyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Fluoropyridin-2-yl)hydrazine, 3-Fluoro-2-hydrazinylpyridine
PC1968-1G
PC1968-5G
PC1968-250MG
APOSPC1968-1GEA
31.9
EUR
APOSPC1968-1G
APOSPC1968-5G
APOSPC1968-250MG
3-Fluoro-2-hydrazinopyridine
3-Fluoro-2-hydrazinopyridine
Formel:
C₅H₆FN₃ Molecular Weight: 127.12 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09998071 CAS-Nummer: 887266-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...