5-Amino-4-fluoro-2-methylphenol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-hydroxy-4-methylaniline, 5-Amino-4-fluoro-o-cresol, 4-Amino-5-fluoro-2-hydroxytoluene, 2-Fluoro-5-hydroxy-p-toluidine

PC1996-1G PC1996-250MG PC1996-5G
APOSPC1996-1GEA 121 EUR
APOSPC1996-1G APOSPC1996-250MG APOSPC1996-5G
5-Amino-4-fluoro-2-methylphenol 98%
5-Amino-4-fluoro-2-methylphenol
Formel: C₇H₈FNO
Molecular Weight: 141,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972656
CAS-Nummer: 122455-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR