Pentafluorophenyl 4-phenylbenzene-1-sulphonate

Lieferant: Apollo Scientific

PC200025-1G PC200025-500MG
APOSPC200025-1GEA 183 EUR
APOSPC200025-1G APOSPC200025-500MG
Pentafluorophenyl 4-phenylbenzene-1-sulphonate
Pentafluorophenyl 4-phenylbenzene-1-sulphonate
Formel: C₁₈H₉F₅O₃S
Molecular Weight: 400,319 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22689145
CAS-Nummer: 1420537-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR