4-(4-Fluorobenzyl)piperidine
Lieferant: Apollo Scientific
Synonyme:
4-(4'-Fluorobenzyl)piperidine
PC200040-1G
PC200040-500MG
PC200040-5G
PC200040-250MG
APOSPC200040-1GEA
165
EUR
APOSPC200040-1G
APOSPC200040-500MG
APOSPC200040-5G
APOSPC200040-250MG
4-(4-Fluorobenzyl)piperidine
4-(4-Fluorobenzyl)piperidine
Formel:
C₁₂H₁₆FN Molecular Weight: 193.261 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03839825 CAS-Nummer: 92822-02-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...