tert-Butyl 4-{[3-fluoro-5-(trifluoromethyl)benzenesulphonyl]methyl}piperidine-1-carboxylate

Lieferant: Apollo Scientific

PC200046-1G PC200046-500MG
APOSPC200046-1GEA 538 EUR
APOSPC200046-1G APOSPC200046-500MG
tert-Butyl 4-{[3-fluoro-5-(trifluoromethyl)benzenesulphonyl]methyl}piperidine-1-carboxylate
tert-Butyl 4-{[3-fluoro-5-(trifluoromethyl)benzenesulphonyl]methyl}piperidine-1-carboxylate
Formel: C₁₈H₂₃F₄NO₄S
Molecular Weight: 425.438 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20482270
CAS-Nummer: 1245784-24-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR