5-Bromo-6-(trifluoromethyl)-1H-indole-3-carbaldehyde

Lieferant: Apollo Scientific

PC200047-1G PC200047-500MG
APOSPC200047-1GEA 634 EUR
APOSPC200047-1G APOSPC200047-500MG
5-Bromo-6-(trifluoromethyl)-1H-indole-3-carbaldehyde
5-Bromo-6-(trifluoromethyl)-1H-indole-3-carbaldehyde
Formel: C₁₀H₅BrF₃NO
Molecular Weight: 292.052 g/mol
Schmelzpunkt: 272…275 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD23136030
CAS-Nummer: 1423757-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR