5-(2-Fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid

Lieferant: Apollo Scientific

PC200060-1G PC200060-500MG
APOSPC200060-1GEA 894 EUR
APOSPC200060-1G APOSPC200060-500MG
5-(2-Fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid
5-(2-Fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid
Formel: C₁₁H₈FNO₂S
Molecular Weight: 237.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08275134
CAS-Nummer: 522646-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR