2-Chloro-4-fluoro-5-iodoanisole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chloro-5-fluoro-4-iodo-2-methoxybenzene, 2-Chloro-4-fluoro-5-iodophenyl methyl ether
PC200065-1G
PC200065-250MG
PC200065-5G
APOSPC200065-1GEA
121
EUR
APOSPC200065-1G
APOSPC200065-250MG
APOSPC200065-5G
2-Chloro-4-fluoro-5-iodoanisole
2-Chloro-4-fluoro-5-iodoanisole
Formel:
C₇H₅ClFIO Molecular Weight: 286.47 g/mol Schmelzpunkt: 104…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22689435 CAS-Nummer: 153122-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...