3-(Trifluoromethyl)pyridine-2-carbonitrile 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Trifluoromethyl)-2-picolinonitrile, 2-Cyano-3-(trifluoromethyl)pyridine, 3-(Trifluoromethyl)picolinonitrile

PC200149-25G
APOSPC200149-25GEA 104 EUR
APOSPC200149-25G
3-(Trifluoromethyl)pyridine-2-carbonitrile 98%
3-(Trifluoromethyl)pyridine-2-carbonitrile
Formel: C₇H₃F₃N₂
Molecular Weight: 172,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02091399
CAS-Nummer: 406933-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR