Potassium [4-(Pyrrolidine-1-carbonyl)phenyl]trifluoroborate

Lieferant: Apollo Scientific

PC200157-1G PC200157-500MG PC200157-5G
APOSPC200157-1GEA 102 EUR
APOSPC200157-1G APOSPC200157-500MG APOSPC200157-5G
Potassium [4-(Pyrrolidine-1-carbonyl)phenyl]trifluoroborate
Potassium [4-(Pyrrolidine-1-carbonyl)phenyl]trifluoroborate
Formel: C₁₁H₁₂BF₃NO·K
Molecular Weight: 281,124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20527094
CAS-Nummer: 1983205-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR