Potassium [4-(benzylamino-1-carbonyl)phenyl]trifluoroborate

Lieferant: Apollo Scientific

PC200159-1G PC200159-5G
APOSPC200159-1GEA 94.5 EUR
APOSPC200159-1G APOSPC200159-5G
Potassium [4-(benzylamino-1-carbonyl)phenyl]trifluoroborate
Potassium [4-(benzylamino-1-carbonyl)phenyl]trifluoroborate
Formel: C₁₄H₁₂BF₃NO·K
Molecular Weight: 317,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977718
CAS-Nummer: 2017555-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR