Potassium 4-butylphenyltrifluoroborate

Lieferant: Apollo Scientific

Synonyme: Potassium (4-butylphenyl)trifluoroboranuide

PC200174-1G PC200174-500MG PC200174-5G
APOSPC200174-1GEA 140 EUR
APOSPC200174-1G APOSPC200174-500MG APOSPC200174-5G
Potassium 4-butylphenyltrifluoroborate
Potassium 4-butylphenyltrifluoroborate
Formel: C₁₀H₁₃BF₃·K
Molecular Weight: 240.115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18861573
CAS-Nummer: 1412414-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR